COM497513176 Combi-Blocks, Inc.

3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02677691
  • Cas Number : 306936-65-2
  • Molecular Weight : 240.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3995

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3995
Availability details for
Quantity
  • Availability
  • Add to Cart