COM497513205 Combi-Blocks, Inc.

6-Bromo-1h-pyrrolo[2,3-b]pyridin-4-amine

  • MDL number MFCD09880154
  • Cas Number : 1000340-72-6
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4125
Availability details for
Quantity
  • Availability
  • Add to Cart