COM497513276 Combi-Blocks, Inc.

Amino-PEG10-amine

  • MDL number MFCD28122962
  • Cas Number : 928292-69-7
  • Molecular Weight : 500.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4687
Availability details for
Quantity
  • Availability
  • Add to Cart