COM497513279 Combi-Blocks, Inc.

3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride

  • MDL number MFCD22392108
  • Cas Number : 1408075-12-6
  • Molecular Weight : 191.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4701
Availability details for
Quantity
  • Availability
  • Add to Cart