COM497513717 Combi-Blocks, Inc.

Sdpd-peg-24-nhs ester

  • MDL number MFCD26793764
  • Molecular Weight : 1439.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7828
Availability details for
Quantity
  • Availability
  • Add to Cart