COM497514015 Combi-Blocks, Inc.

1-Acetylpiperidin-3-yl acetate

  • MDL number MFCD23378590
  • Cas Number : 42366-59-6
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0021
Availability details for
Quantity
  • Availability
  • Add to Cart