COM497514123 Combi-Blocks, Inc.

4-(4-Ethylpiperazin-1-yl)aniline hcl

  • MDL number MFCD28133448
  • Cas Number : 1172518-57-8
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0251

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0251
Availability details for
Quantity
  • Availability
  • Add to Cart