COM497514243 Combi-Blocks, Inc.

1-(2-Ethoxybenzyl)-2-methylhydrazine

  • MDL number MFCD28053739
  • Cas Number : 1392879-03-6
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0523
Availability details for
Quantity
  • Availability
  • Add to Cart