COM497514532 Combi-Blocks, Inc.

3-Aminoquinoline-7-carboxylic acid

  • MDL number MFCD28133486
  • Cas Number : 1824051-45-7
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1170
Availability details for
Quantity
  • Availability
  • Add to Cart