COM497514599 Combi-Blocks, Inc.

Benzyl 2-(benzyloxycarbonylamino)-3-(4-iodophenyl)propanoate

  • MDL number MFCD23378606
  • Cas Number : 150334-32-0
  • Molecular Weight : 515.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1332
Availability details for
Quantity
  • Availability
  • Add to Cart