COM497514650 Combi-Blocks, Inc.

2-(1-Methyl-1h-indol-3-yl)propanenitrile

  • MDL number MFCD20039889
  • Cas Number : 176688-64-5
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1463
Availability details for
Quantity
  • Availability
  • Add to Cart