COM497514720 Combi-Blocks, Inc.

5-Fluoro-4-methoxy-2-nitroaniline

  • MDL number MFCD12405136
  • Cas Number : 446-20-8
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1621
Availability details for
Quantity
  • Availability
  • Add to Cart