COM497514857 Combi-Blocks, Inc.

Methyl 3-bromo-4-(tert-butoxycarbonyl(methyl)amino)benzoate

  • MDL number MFCD28359627
  • Cas Number : 1951440-95-1
  • Molecular Weight : 344.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1921
Availability details for
Quantity
  • Availability
  • Add to Cart