COM497514928 Combi-Blocks, Inc.

Psi-7977

  • MDL number MFCD18782704
  • Cas Number : 1064684-44-1
  • Molecular Weight : 529.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2117
Availability details for
Quantity
  • Availability
  • Add to Cart