COM497515021 Combi-Blocks, Inc.

2-Chloro-n-(cyclopropylmethyl)-n-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)acetamide

  • MDL number MFCD09909356
  • Cas Number : 1951441-02-3
  • Molecular Weight : 389.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2333
Availability details for
Quantity
  • Availability
  • Add to Cart