COM497515045 Combi-Blocks, Inc.

(4-Chloro-3-fluorophenyl)acetic acid ethyl ester

  • MDL number MFCD11045166
  • Cas Number : 1256479-12-5
  • Molecular Weight : 216.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2399
Availability details for
Quantity
  • Availability
  • Add to Cart