COM497515081 Combi-Blocks, Inc.

4-Chloro-2,3-dihydro-1h-pyrrolo[3,2-c]pyridine hydrochloride

  • MDL number MFCD14586358
  • Cas Number : 1241725-93-8
  • Molecular Weight : 191.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2475

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2475
Availability details for
Quantity
  • Availability
  • Add to Cart