COM497515114 Combi-Blocks, Inc.

(R)-tert-Butyl 3-carbamoylpiperazine-1-carboxylate

  • MDL number MFCD03094732
  • Cas Number : 170164-46-2
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2554
Availability details for
Quantity
  • Availability
  • Add to Cart