COM497515117 Combi-Blocks, Inc.

3-Amino-6-ethyl-2-thioxo-2,3-dihydro-1h-pyrimidin-4-one

  • MDL number MFCD14585459
  • Cas Number : 1206969-66-5
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2559

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2559
Availability details for
Quantity
  • Availability
  • Add to Cart