COM497515196 Combi-Blocks, Inc.

(4-Bromothiazol-2-yl)methanamine hcl

  • MDL number MFCD09878587
  • Cas Number : 1207175-70-9
  • Molecular Weight : 229.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2753
Availability details for
Quantity
  • Availability
  • Add to Cart