COM497515325 Combi-Blocks, Inc.

Carboxymethanesulfinyl-acetic acid

  • MDL number MFCD03042673
  • Cas Number : 33388-14-6
  • Molecular Weight : 166.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3029
Availability details for
Quantity
  • Availability
  • Add to Cart