COM497515328 Combi-Blocks, Inc.

(2R,5R)-2,5-Dimethylpyrrolidine

  • MDL number MFCD01863020
  • Cas Number : 62617-70-3
  • Molecular Weight : 99.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3038
Availability details for
Quantity
  • Availability
  • Add to Cart