COM497515390 Combi-Blocks, Inc.

2-Amino-2,4-dimethylpentanenitrile

  • MDL number MFCD01938367
  • Cas Number : 26842-43-3
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3180
Availability details for
Quantity
  • Availability
  • Add to Cart