COM497515450 Combi-Blocks, Inc.

Gambogic acid

  • MDL number MFCD16878985
  • Cas Number : 2752-65-0
  • Molecular Weight : 628.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3313
Availability details for
Quantity
  • Availability
  • Add to Cart