COM497516047 Combi-Blocks, Inc.

1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-l-proline hydrate

  • MDL number MFCD07809564
  • Cas Number : 64838-55-7
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4666
Availability details for
Quantity
  • Availability
  • Add to Cart