COM497516083 Combi-Blocks, Inc.

1-(3-Fluoro-phenyl)-piperazine dihydrochloride

  • MDL number MFCD03540953
  • Cas Number : 3801-89-6
  • Molecular Weight : 253.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4754
Availability details for
Quantity
  • Availability
  • Add to Cart