COM497516281 Combi-Blocks, Inc.

Methyl 4-(2-pyridinyl)benzoate

  • MDL number MFCD00110081
  • Cas Number : 98061-21-3
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5207
Availability details for
Quantity
  • Availability
  • Add to Cart