COM497516413 Combi-Blocks, Inc.

1-Bromo-3-chloro-2,4,5-trifluorobenzene

  • MDL number MFCD26960578
  • Cas Number : 152840-71-6
  • Molecular Weight : 245.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5486

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5486
Availability details for
Quantity
  • Availability
  • Add to Cart