COM497516480 Combi-Blocks, Inc.

2-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)ethanone

  • MDL number MFCD18452152
  • Cas Number : 94171-11-6
  • Molecular Weight : 299.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5639
Availability details for
Quantity
  • Availability
  • Add to Cart