COM497516720 Combi-Blocks, Inc.

1-Aminocyclopentane-1-carboxamide hydrochloride

  • MDL number MFCD18838831
  • Cas Number : 17704-76-6
  • Molecular Weight : 164.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6209
Availability details for
Quantity
  • Availability
  • Add to Cart