COM497516793 Combi-Blocks, Inc.

3-Cyano-5-methylbenzoic acid

  • MDL number MFCD11007822
  • Cas Number : 78621-81-5
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6407
Availability details for
Quantity
  • Availability
  • Add to Cart