COM497516824 Combi-Blocks, Inc.

3-Chloroquinolin-5-amine

  • MDL number MFCD19690251
  • Cas Number : 873969-81-4
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6500
Availability details for
Quantity
  • Availability
  • Add to Cart