COM497516833 Combi-Blocks, Inc.

3-Aminoquinoline-5-carboxylic acid

  • MDL number MFCD26710349
  • Cas Number : 1539636-62-8
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6521
Availability details for
Quantity
  • Availability
  • Add to Cart