COM497516934 Combi-Blocks, Inc.

8-(2,6-Difluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane

  • MDL number MFCD23378771
  • Cas Number : 1332356-41-8
  • Molecular Weight : 300.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-6746
Availability details for
Quantity
  • Availability
  • Add to Cart