COM497517028 Combi-Blocks, Inc.

tert-Butyl 3 hydroxy-4-nitrobenzoate

  • MDL number MFCD08703419
  • Cas Number : 123330-86-9
  • Molecular Weight : 239.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7003
Availability details for
Quantity
  • Availability
  • Add to Cart