COM497517042 Combi-Blocks, Inc.

N-(4-Methoxy-3-methylphenyl)acetamide

  • MDL number MFCD00128802
  • Cas Number : 31910-25-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7031
Availability details for
Quantity
  • Availability
  • Add to Cart