COM497517054 Combi-Blocks, Inc.

N-Methyl-dl-aspartic acid

  • MDL number MFCD00065918
  • Cas Number : 17833-53-3
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7053
Availability details for
Quantity
  • Availability
  • Add to Cart