COM497517064 Combi-Blocks, Inc.

2-((S)-1-(7-Methyl-2-morpholino-4-oxo-4h-pyrido[1,2-a]pyrimidin-9-yl)ethylamino)benzoic acid

  • MDL number MFCD28053500
  • Cas Number : 1173900-37-2
  • Molecular Weight : 408.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7076
Availability details for
Quantity
  • Availability
  • Add to Cart