COM497517107 Combi-Blocks, Inc.

Methyl 2-amino-5-chloro-3-nitrobenzoate

  • MDL number MFCD02068568
  • Cas Number : 84228-49-9
  • Molecular Weight : 230.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7192
Availability details for
Quantity
  • Availability
  • Add to Cart