COM497517432 Combi-Blocks, Inc.

1-[(1S)-1-(4-chloro-3-fluorophenyl)-2-hydroxyethyl]-4-{2-[(2-methylpyrazol-3-yl)amino]pyrimidin-4-yl}pyridin-2-one

  • MDL number MFCD28143918
  • Cas Number : 1453848-26-4
  • Molecular Weight : 440.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7929
Availability details for
Quantity
  • Availability
  • Add to Cart