COM497517498 Combi-Blocks, Inc.

3-(3-Methoxyphenyl)pyrrolidine

  • MDL number MFCD09027613
  • Cas Number : 38175-35-8
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8099
Availability details for
Quantity
  • Availability
  • Add to Cart