COM497517542 Combi-Blocks, Inc.

5-Fluoro-1h-indole-2-carbaldehyde

  • MDL number MFCD06800739
  • Cas Number : 220943-23-7
  • Molecular Weight : 163.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8195
Availability details for
Quantity
  • Availability
  • Add to Cart