COM497517553 Combi-Blocks, Inc.

4-(2-Fluorophenyl)butanenitrile

  • MDL number MFCD09744513
  • Cas Number : 143654-61-9
  • Molecular Weight : 163.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8214
Availability details for
Quantity
  • Availability
  • Add to Cart