COM497517693 Combi-Blocks, Inc.

(5-(4-Fluoro-2-(trifluoromethyl)phenyl)-1,3,4-thiadiazol-2-yl)methyl acetate

  • MDL number MFCD19689402
  • Cas Number : 1951438-92-8
  • Molecular Weight : 320.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8522
Availability details for
Quantity
  • Availability
  • Add to Cart