COM497517825 Combi-Blocks, Inc.

tert-Butyl 2-(piperidin-4-yl)pyrrolidine-1-carboxylate

  • MDL number MFCD09702201
  • Cas Number : 929974-12-9
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8873
Availability details for
Quantity
  • Availability
  • Add to Cart