COM497517920 Combi-Blocks, Inc.

Bis(benzyltriphenylphosphonium)peroxydisulfate

  • MDL number MFCD20133824
  • Molecular Weight : 898.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9116
Availability details for
Quantity
  • Availability
  • Add to Cart