COM497517922 Combi-Blocks, Inc.

4-Hydroxy-7-methoxy-3-quinolinecarbonitrile

  • MDL number MFCD13181181
  • Cas Number : 2305-69-3
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9120
Availability details for
Quantity
  • Availability
  • Add to Cart