COM497518129 Combi-Blocks, Inc.

tert-Butyl 4-nitro-1h-imidazole-1-carboxylate

  • MDL number MFCD20039783
  • Cas Number : 716316-20-0
  • Molecular Weight : 213.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9570
Availability details for
Quantity
  • Availability
  • Add to Cart