COM497518203 Combi-Blocks, Inc.

tert-Butyl 2-(4-fluorophenyl)acetate

  • MDL number MFCD08310687
  • Cas Number : 68825-45-6
  • Molecular Weight : 210.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9733
Availability details for
Quantity
  • Availability
  • Add to Cart