COM497518225 Combi-Blocks, Inc.

5-(tert-Butyldimethylsilyloxy)-4,4-dimethyl-3-oxopentanenitrile

  • MDL number MFCD26399496
  • Cas Number : 1229454-44-7
  • Molecular Weight : 255.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9789
Availability details for
Quantity
  • Availability
  • Add to Cart